N-(Cyclopentylcarbonyl)alanine

Supplier: Matrix Scientific

049350-500MG 049350-2.5G
101887-304EA 364.62 USD
101887-304 101887-306
N-(Cyclopentylcarbonyl)alanine
N-(Cyclopentylcarbonyl)alanine

Hazard: Irritant

Formula: C₉H₁₅NO₃
MDL Number: MFCD09836859
CAS Number: 252678-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR