2-[(Cyclopentylcarbonyl)amino]acetic acid

Supplier: Matrix Scientific

049349-500MG 049349-2.5G
101887-300EA 364.62 USD
101887-300 101887-302
2-[(Cyclopentylcarbonyl)amino]acetic acid
2-[(Cyclopentylcarbonyl)amino]acetic acid

Hazard: Irritant

Formula: C₈H₁₃NO₃
MDL Number: MFCD08445065
CAS Number: 82291-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR