3-Pyrrolidinyl cyclopentanecarboxylate hydrochloride

Supplier: Matrix Scientific

049346-500MG 049346-2.5G
101887-288EA 364.62 USD
101887-288 101887-290
3-Pyrrolidinyl cyclopentanecarboxylate hydrochloride
3-Pyrrolidinyl cyclopentanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClNO₂
MDL Number: MFCD13559837
CAS Number: 1219980-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR