3-Pyrrolidinylmethyl cyclopentanecarboxylate hydrochloride

Supplier: Matrix Scientific

049345-500MG 049345-2.5G
101887-284EA 364.62 USD
101887-284 101887-286
3-Pyrrolidinylmethyl cyclopentanecarboxylate hydrochloride
3-Pyrrolidinylmethyl cyclopentanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₀ClNO₂
MDL Number: MFCD13559836
CAS Number: 1220038-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR