2-(2-Piperidinyl)ethyl cyclopentanecarboxylate hydrochloride

Supplier: Matrix Scientific

049344-500MG 049344-2.5G
101887-280EA 364.62 USD
101887-280 101887-282
2-(2-Piperidinyl)ethyl cyclopentanecarboxylate hydrochloride
2-(2-Piperidinyl)ethyl cyclopentanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₄ClNO₂
MDL Number: MFCD13559835
CAS Number: 1220027-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR