2-(4-Piperidinyl)ethyl cyclopentanecarboxylate hydrochloride

Supplier: Matrix Scientific

049342-500MG 049342-2.5G
101887-272EA 364.62 USD
101887-272 101887-274
2-(4-Piperidinyl)ethyl cyclopentanecarboxylate hydrochloride
2-(4-Piperidinyl)ethyl cyclopentanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₄ClNO₂
MDL Number: MFCD13559833
CAS Number: 1220021-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR