3-Piperidinylmethyl cyclopentanecarboxylate hydrochloride

Supplier: Matrix Scientific

049340-500MG 049340-2.5G
101887-264EA 364.62 USD
101887-264 101887-266
3-Piperidinylmethyl cyclopentanecarboxylate hydrochloride
3-Piperidinylmethyl cyclopentanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₂ClNO₂
MDL Number: MFCD13559831
CAS Number: 1220021-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR