4-Piperidinyl cyclopentanecarboxylate hydrochloride

Supplier: Matrix Scientific

049338-500MG 049338-2.5G
101887-256EA 364.62 USD
101887-256 101887-258
4-Piperidinyl cyclopentanecarboxylate hydrochloride
4-Piperidinyl cyclopentanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₀ClNO₂
MDL Number: MFCD13559829
CAS Number: 1219948-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR