3-Piperidinyl cyclopentanecarboxylate hydrochloride
Supplier: Matrix Scientific
049337-500MG
049337-2.5G
101887-252EA
364.62
USD
101887-252
101887-254
3-Piperidinyl cyclopentanecarboxylate hydrochloride
3-Piperidinyl cyclopentanecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₀ClNO₂ MW: 233.74 g/mol |
MDL Number:
MFCD13559828 CAS Number: 1219972-60-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...