N-(Cyclobutylcarbonyl)-2-methylalanine

Supplier: Matrix Scientific

Synonyms: 2-(Cyclobutanecarboxamido)-2-methylpropanoic acid

049334-500MG 049334-2.5G
101887-240EA 364.62 USD
101887-240 101887-242
N-(Cyclobutylcarbonyl)-2-methylalanine
N-(Cyclobutylcarbonyl)-2-methylalanine

Hazard: Irritant

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
MDL Number: MFCD13559827
CAS Number: 1219960-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR