1-(Cyclobutylcarbonyl)-3-piperidinecarboxylic acid

Supplier: Matrix Scientific

049333-500MG 049333-2.5G
101887-236EA 364.62 USD
101887-236 101887-238
1-(Cyclobutylcarbonyl)-3-piperidinecarboxylic acid
1-(Cyclobutylcarbonyl)-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₇NO₃
MDL Number: MFCD09043166
CAS Number: 926214-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR