1-(Cyclobutylcarbonyl)-2-piperidinecarboxylic acid

Supplier: Matrix Scientific

049332-500MG 049332-2.5G
101887-232EA 364.62 USD
101887-232 101887-234
1-(Cyclobutylcarbonyl)-2-piperidinecarboxylic acid
1-(Cyclobutylcarbonyl)-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₇NO₃
MDL Number: MFCD09050725
CAS Number: 1101762-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR