1-(Cyclobutylcarbonyl)proline

Supplier: Matrix Scientific

049331-500MG 049331-2.5G
101887-228EA 364.62 USD
101887-228 101887-230
1-(Cyclobutylcarbonyl)proline
1-(Cyclobutylcarbonyl)proline

Hazard: Irritant

Formula: C₁₀H₁₅NO₃
MDL Number: MFCD09048570
CAS Number: 1101745-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR