N-(Cyclobutylcarbonyl)alanine

Supplier: Matrix Scientific

049330-500MG 049330-2.5G
101887-224EA 364.62 USD
101887-224 101887-226
N-(Cyclobutylcarbonyl)alanine
N-(Cyclobutylcarbonyl)alanine

Hazard: Irritant

Formula: C₈H₁₃NO₃
MDL Number: MFCD09047833
CAS Number: 118636-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR