2-[(Cyclobutylcarbonyl)amino]acetic acid

Supplier: Matrix Scientific

049329-500MG 049329-2.5G
101887-220EA 364.62 USD
101887-220 101887-222
2-[(Cyclobutylcarbonyl)amino]acetic acid
2-[(Cyclobutylcarbonyl)amino]acetic acid

Hazard: Irritant

Formula: C₇H₁₁NO₃
MDL Number: MFCD09048424
CAS Number: 604790-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR