N-(4-Piperidinyl)cyclobutanecarboxamide hydrochloride
Supplier: Matrix Scientific
049328-500MG
049328-2.5G
101887-216EA
364.62
USD
101887-216
101887-218
N-(4-Piperidinyl)cyclobutanecarboxamide hydrochloride
N-(4-Piperidinyl)cyclobutanecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₉ClN₂O |
MDL Number:
MFCD13559826 CAS Number: 1219948-63-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...