Methyl (2S,4S)-4-[(cyclobutylcarbonyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

049327-500MG 049327-2.5G
101887-212EA 364.62 USD
101887-212 101887-214
Methyl (2S,4S)-4-[(cyclobutylcarbonyl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[(cyclobutylcarbonyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈ClNO₄
MDL Number: MFCD13559825
CAS Number: 1354487-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR