3-Pyrrolidinyl cyclobutanecarboxylate hydrochloride

Supplier: Matrix Scientific

049326-500MG 049326-2.5G
101887-208EA 364.62 USD
101887-208 101887-210
3-Pyrrolidinyl cyclobutanecarboxylate hydrochloride
3-Pyrrolidinyl cyclobutanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₉H₁₆ClNO₂
MDL Number: MFCD13559824
CAS Number: 1220032-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR