3-Pyrrolidinylmethyl cyclobutanecarboxylate hydrochloride

Supplier: Matrix Scientific

049325-500MG 049325-2.5G
101887-204EA 364.62 USD
101887-204 101887-206
3-Pyrrolidinylmethyl cyclobutanecarboxylate hydrochloride
3-Pyrrolidinylmethyl cyclobutanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClNO₂
MDL Number: MFCD13559823
CAS Number: 1220021-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR