2-(3-Piperidinyl)ethyl cyclobutanecarboxylate hydrochloride

Supplier: Matrix Scientific

049323-500MG 049323-2.5G
101887-196EA 364.62 USD
101887-196 101887-198
2-(3-Piperidinyl)ethyl cyclobutanecarboxylate hydrochloride
2-(3-Piperidinyl)ethyl cyclobutanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₂ClNO₂
MDL Number: MFCD13559821
CAS Number: 1219972-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR