4-Piperidinylmethyl cyclobutanecarboxylate hydrochloride

Supplier: Matrix Scientific

049321-500MG 049321-2.5G
101887-188EA 364.62 USD
101887-188 101887-190
4-Piperidinylmethyl cyclobutanecarboxylate hydrochloride
4-Piperidinylmethyl cyclobutanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₀ClNO₂
MDL Number: MFCD13559819
CAS Number: 1219980-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR