3-Piperidinylmethyl cyclobutanecarboxylate hydrochloride

Supplier: Matrix Scientific

049320-500MG 049320-2.5G
101887-184EA 364.62 USD
101887-184 101887-186
3-Piperidinylmethyl cyclobutanecarboxylate hydrochloride
3-Piperidinylmethyl cyclobutanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₀ClNO₂
MDL Number: MFCD13559818
CAS Number: 1219960-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR