3-Azetidinyl cyclobutanecarboxylate
Supplier: Matrix Scientific
049319-500MG
049319-2.5G
101887-180EA
364.62
USD
101887-180
101887-182
3-Azetidinyl cyclobutanecarboxylate
3-Azetidinyl cyclobutanecarboxylate
Hazard: Irritant
Formula:
C₈H₁₃NO₂ MW: 155.2 g/mol |
MDL Number:
MFCD13559817 CAS Number: 1220027-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...