2-[(Cyclopropylcarbonyl)(methyl)amino]acetic acid

Supplier: Matrix Scientific

049315-500MG 049315-2.5G
101887-164EA 364.62 USD
101887-164 101887-166
2-[(Cyclopropylcarbonyl)(methyl)amino]acetic acid
2-[(Cyclopropylcarbonyl)(methyl)amino]acetic acid

Hazard: Irritant

Formula: C₇H₁₁NO₃
MDL Number: MFCD09050622
CAS Number: 926213-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR