N-(Cyclopropylcarbonyl)-2-methylalanine

Supplier: Matrix Scientific

049314-500MG 049314-2.5G
101887-160EA 364.62 USD
101887-160 101887-162
N-(Cyclopropylcarbonyl)-2-methylalanine
N-(Cyclopropylcarbonyl)-2-methylalanine

Hazard: Irritant

Formula: C₈H₁₃NO₃
MDL Number: MFCD13559814
CAS Number: 215036-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR