1-(Cyclopropylcarbonyl)proline

Supplier: Matrix Scientific

Synonyms: (S)-1-(Cyclopropanecarbonyl)pyrrolidine-2-carboxylic acid

049311-500MG 049311-2.5G
101887-148EA 364.62 USD
101887-148 101887-150
1-(Cyclopropylcarbonyl)proline
1-(Cyclopropylcarbonyl)proline

Hazard: Irritant

Formula: C₉H₁₃NO₃
MW: 183.21 g/mol
MDL Number: MFCD09045460
CAS Number: 148706-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR