N-(Cyclopropylcarbonyl)alanine

Supplier: Matrix Scientific

049310-500MG 049310-2.5G
101887-144EA 364.62 USD
101887-144 101887-146
N-(Cyclopropylcarbonyl)alanine
N-(Cyclopropylcarbonyl)alanine

Hazard: Irritant

Formula: C₇H₁₁NO₃
MDL Number: MFCD09049279
CAS Number: 1397008-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR