2-[(Cyclopropylcarbonyl)amino]acetic acid

Supplier: Matrix Scientific

049309-500MG 049309-2.5G
101887-140EA 364.62 USD
101887-140 101887-142
2-[(Cyclopropylcarbonyl)amino]acetic acid
2-[(Cyclopropylcarbonyl)amino]acetic acid

Hazard: Irritant

Formula: C₆H₉NO₃
MDL Number: MFCD01724840
CAS Number: 64513-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR