Methyl (2S,4S)-4-[(cyclopropylcarbonyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

049307-500MG 049307-2.5G
101887-132EA 364.62 USD
101887-132 101887-134
Methyl (2S,4S)-4-[(cyclopropylcarbonyl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[(cyclopropylcarbonyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆ClNO₄
MDL Number: MFCD13559812
CAS Number: 1354486-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR