3-Pyrrolidinyl cyclopropanecarboxylate hydrochloride
Supplier: Matrix Scientific
049306-500MG
049306-2.5G
101887-128EA
364.62
USD
101887-128
101887-130
3-Pyrrolidinyl cyclopropanecarboxylate hydrochloride
3-Pyrrolidinyl cyclopropanecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₈H₁₄ClNO₂ |
MDL Number:
MFCD13559811 CAS Number: 1219960-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...