3-Pyrrolidinylmethyl cyclopropanecarboxylate hydrochloride

Supplier: Matrix Scientific

049305-500MG 049305-2.5G
101887-124EA 364.62 USD
101887-124 101887-126
3-Pyrrolidinylmethyl cyclopropanecarboxylate hydrochloride
3-Pyrrolidinylmethyl cyclopropanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₉H₁₆ClNO₂
MDL Number: MFCD13559810
CAS Number: 1219980-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR