2-(3-Piperidinyl)ethyl cyclopropanecarboxylate hydrochloride
Supplier: Matrix Scientific
049303-500MG
049303-2.5G
101887-116EA
364.62
USD
101887-116
101887-118
2-(3-Piperidinyl)ethyl cyclopropanecarboxylate hydrochloride
2-(3-Piperidinyl)ethyl cyclopropanecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₀ClNO₂ |
MDL Number:
MFCD13559808 CAS Number: 1219948-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...