2-(4-Piperidinyl)ethyl cyclopropanecarboxylate hydrochloride
Supplier: Matrix Scientific
049302-500MG
049302-2.5G
101887-112EA
364.62
USD
101887-112
101887-114
2-(4-Piperidinyl)ethyl cyclopropanecarboxylate hydrochloride
2-(4-Piperidinyl)ethyl cyclopropanecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₀ClNO₂ |
MDL Number:
MFCD13559807 CAS Number: 1220038-23-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...