4-Piperidinylmethyl cyclopropanecarboxylate hydrochloride

Supplier: Matrix Scientific

049301-500MG 049301-2.5G
101887-108EA 364.62 USD
101887-108 101887-110
4-Piperidinylmethyl cyclopropanecarboxylate hydrochloride
4-Piperidinylmethyl cyclopropanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClNO₂
MDL Number: MFCD13559806
CAS Number: 1219968-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR