N-(3-Methylbutanoyl)-β-alanine

Supplier: Matrix Scientific

049296-500MG 049296-2.5G
101887-088EA 364.62 USD
101887-088 101887-090
N-(3-Methylbutanoyl)-β-alanine
N-(3-Methylbutanoyl)-beta-alanine

Hazard: Irritant

Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
MDL Number: MFCD08445040
CAS Number: 923249-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR