1-(3-Methylbutanoyl)proline
Supplier: Matrix Scientific
049291-500MG
049291-2.5G
101887-068EA
364.62
USD
101887-068
101887-070
1-(3-Methylbutanoyl)proline
1-(3-Methylbutanoyl)proline
Hazard: Irritant
Formula:
C₁₀H₁₇NO₃ |
MDL Number:
MFCD08442897 CAS Number: 60804-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...