N-Isovalerylglycine

Supplier: Matrix Scientific

Synonyms: 2-(3-Methylbutanamido)acetic acid, N-Isovaleroylglycine

049289-500MG 049289-2.5G
101887-060EA 115.38 USD
101887-060 101887-062
N-Isovalerylglycine
N-Isovalerylglycine

Hazard: Irritant

Formula: C₇H₁₃NO₃
MW: 159.19 g/mol
MDL Number: MFCD00059629
CAS Number: 16284-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR