3-Methyl-N-(4-piperidinyl)butanamide hydrochloride

Supplier: Matrix Scientific

049288-500MG 049288-2.5G
101887-056EA 364.62 USD
101887-056 101887-058
3-Methyl-N-(4-piperidinyl)butanamide hydrochloride
3-Methyl-N-(4-piperidinyl)butanamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD13559800
CAS Number: 1220027-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR