3-Pyrrolidinyl-3-methylbutanoate hydrochloride

Supplier: Matrix Scientific

049286-500MG 049286-2.5G
101887-048EA 364.62 USD
101887-048 101887-050
3-Pyrrolidinyl-3-methylbutanoate hydrochloride
3-Pyrrolidinyl-3-methylbutanoate hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₂
MDL Number: MFCD13559798
CAS Number: 1219960-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR