3-Pyrrolidinylmethyl-3-methylbutanoate hydrochloride

Supplier: Matrix Scientific

049285-500MG 049285-2.5G
101887-044EA 364.62 USD
101887-044 101887-046
3-Pyrrolidinylmethyl-3-methylbutanoate hydrochloride
3-Pyrrolidinylmethyl-3-methylbutanoate hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO₂
MDL Number: MFCD13559797
CAS Number: 1220038-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR