2-(2-Piperidinyl)ethyl-3-methylbutanoate hydrochloride
Supplier: Matrix Scientific
049284-500MG
049284-2.5G
101887-040EA
364.62
USD
101887-040
101887-042
2-(2-Piperidinyl)ethyl-3-methylbutanoate hydrochloride
2-(2-Piperidinyl)ethyl-3-methylbutanoate hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₄ClNO₂ |
MDL Number:
MFCD13559796 CAS Number: 1220021-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...