2-(3-Piperidinyl)ethyl-3-methylbutanoate hydrochloride

Supplier: Matrix Scientific

049283-500MG 049283-2.5G
101887-036EA 364.62 USD
101887-036 101887-038
2-(3-Piperidinyl)ethyl-3-methylbutanoate hydrochloride
2-(3-Piperidinyl)ethyl-3-methylbutanoate hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₄ClNO₂
MDL Number: MFCD13559795
CAS Number: 1219960-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR