2-(4-Piperidinyl)ethyl-3-methylbutanoate hydrochloride

Supplier: Matrix Scientific

049282-500MG 049282-2.5G
101887-032EA 364.62 USD
101887-032 101887-034
2-(4-Piperidinyl)ethyl-3-methylbutanoate hydrochloride
2-(4-Piperidinyl)ethyl-3-methylbutanoate hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₄ClNO₂
MDL Number: MFCD13559794
CAS Number: 1219948-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR