4-Piperidinylmethyl-3-methylbutanoate hydrochloride

Supplier: Matrix Scientific

049281-500MG 049281-2.5G
101887-028EA 364.62 USD
101887-028 101887-030
4-Piperidinylmethyl-3-methylbutanoate hydrochloride
4-Piperidinylmethyl-3-methylbutanoate hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₂ClNO₂
MW: 235.76 g/mol
MDL Number: MFCD13559793
CAS Number: 1219979-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR