3-Azetidinyl-3-methylbutanoate

Supplier: Matrix Scientific

049279-500MG 049279-2.5G
101887-020EA 364.62 USD
101887-020 101887-022
3-Azetidinyl-3-methylbutanoate
3-Azetidinyl-3-methylbutanoate

Hazard: Irritant

Formula: C₈H₁₅NO₂
MDL Number: MFCD13559791
CAS Number: 1219948-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR