N-Isobutyryl-β-alanine

Supplier: Matrix Scientific

049276-500MG 049276-2.5G
101887-008EA 364.62 USD
101887-008 101887-010
N-Isobutyryl-β-alanine
N-Isobutyryl-beta-alanine

Hazard: Irritant

Formula: C₇H₁₃NO₃
MDL Number: MFCD09044885
CAS Number: 126424-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR