2-[Isobutyryl(methyl)amino]acetic acid

Supplier: Matrix Scientific

049275-500MG 049275-2.5G
101887-004EA 364.62 USD
101887-004 101887-006
2-[Isobutyryl(methyl)amino]acetic acid
2-[Isobutyryl(methyl)amino]acetic acid

Hazard: Irritant

Formula: C₇H₁₃NO₃
MDL Number: MFCD09045779
CAS Number: 155256-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR