N-Isobutyryl-2-methylalanine

Supplier: Matrix Scientific

049274-500MG 049274-2.5G
101887-000EA 364.62 USD
101887-000 101887-002
N-Isobutyryl-2-methylalanine
N-Isobutyryl-2-methylalanine

Hazard: Irritant

Formula: C₈H₁₅NO₃
MDL Number: MFCD13559788
CAS Number: 118728-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR