1-Isobutyryl-4-piperidinecarboxylic acid

Supplier: Matrix Scientific

049273-500MG 049273-2.5G
101886-996EA 364.62 USD
101886-996 101886-998
1-Isobutyryl-4-piperidinecarboxylic acid
1-Isobutyryl-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₃
MDL Number: MFCD09043253
CAS Number: 330985-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR