1-Isobutyryl-2-piperidinecarboxylic acid
Supplier: Matrix Scientific
049271-500MG
049271-2.5G
101886-988EA
364.62
USD
101886-988
101886-990
1-Isobutyryl-2-piperidinecarboxylic acid
1-Isobutyryl-2-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₇NO₃ |
MDL Number:
MFCD09048715 CAS Number: 1103336-91-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...